missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ PTFE Magnetic Stirring Bars, Cylindrical Shape
Manufactured from PTFE. BRAND™ PTFE Magnetic Stirring Bars, Cylindrical Shape are suitable for universal applications. Rounded, smooth surface provides effective stirring action.
Brand: BRAND™ 137108
33.79 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Manufactured from PTFE
- Rounded, smooth surface provides effective stirring action.
- Suitable for universal applications.
Specifications
3 mm | |
PTFE | |
Cylinder |
10 mm | |
10/Pk. |