missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ PTFE Magnetic Stirring Bars, Cylindrical Shape

Manufactured from PTFE. BRAND™ PTFE Magnetic Stirring Bars, Cylindrical Shape are suitable for universal applications. Rounded, smooth surface provides effective stirring action.

Brand:  BRAND™ 137105

17.04 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.



Code : A0

Additional Details : Weight : 0.00810kg

 View more versions of this product

Product Code. 11854140

  • £31.95 / Pack of 10
In Stock
to see stock.
Add to basket

Description

Description

  • Manufactured from PTFE
  • Rounded, smooth surface provides effective stirring action.
  • Suitable for universal applications.
Specifications

Specifications

4.5 mm
PTFE
Cylinder
10/Pk.
12 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers