Learn More
BRAND™ PTFE Magnetic Stirring Bars, Cylindrical Shape
Manufactured from PTFE. BRAND™ PTFE Magnetic Stirring Bars, Cylindrical Shape are suitable for universal applications. Rounded, smooth surface provides effective stirring action.
Brand:  BRAND™ 137104
26.45 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 12020922
  • £38.50 / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from PTFE
  • Rounded, smooth surface provides effective stirring action.
  • Suitable for universal applications.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PTFE
8 mm
10/Pk.
Magnetic Stir Bar
3 mm
PTFE
Cylinder