Learn More
Kartell™ Plastilab™ All-Purpose Cylindrical Stirring Bars
Features rounded shape with smooth walls.
Brand: Kartell™ 0075100
This item is not returnable.
View return policy
Description
Features rounded shape with smooth walls.
- PTFE-coated magnet
- Magnetic field causes stirring bar to spin very quickly
- Non-stick, non-reactive and hydrophobic
- Highly resistant to thermo-shock
- Suitable for foodstuff
Specifications
Magnet PTFE Coated | |
14.80 mm | |
Cylinder, Sphere |
4.57 mm | |
10/Pk. | |
Stir Bar |