Learn More
Cymoxanil, Honeywell Fluka™
Brand: Honeywell Fluka R1785-100MG
Additional Details : CAS Number : 57966-95-7 Weight : 0.10000kg
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Cymoxanil | |
≤ 100% | |
2214018 | |
198.18 g/mol | |
161°C | |
100 mg |
57966-95-7 | |
C7H10N4O3 | |
2-cyano-N-[(ethylamino)carbonyl]-2-(methoxyimino)acetamide | |
1.310 g/cm3 (20°C) | |
Colorless |