missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP3A43 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564277
This item is not returnable.
View return policy
Description
Immunogen sequence: FLGTILFYLR GLWNFDRECN EKYGE Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 60%.
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme has a low level of testosterone hydroxylase activity. Although it bears homology to some drug-metabolizing cytochrome P450s, it is unknown whether the enzyme is also involved in xenobiotic metabolism. This gene is part of a cluster of cytochrome P450 genes on chromosome 7q21.1. Alternate splicing of this gene results in three transcript variants encoding different isoforms.Specifications
CYP3A43 | |
Polyclonal | |
Unconjugated | |
CYP3A43 | |
CYP3A43; Cytochrome P450 3A43; cytochrome P450 family 3 subfamily A member 43; cytochrome P450, family 3, subfamily A, polypeptide 43; cytochrome P450, subfamily IIIA, polypeptide 43 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64816 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HB55 | |
CYP3A43 | |
Recombinant protein corresponding to Human CYP3A43 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only