missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cypermethrin, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Density | 1.1200g/mL |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (Mixture of isomers) (HPLC) |
Quantity | 5 g |
Specific Gravity | 1.12 |
Solubility Information | Solubility in water: insoluble. |
Formula Weight | 416.3 |
Percent Purity | 96% |
Chemical Name or Material | Cypermethrin |
Safety and Handling


Product Identifier
- Cypermethrin
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Specific target organ toxicity Category 3
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H301-Toxic if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cypermethrin, 96%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction