Learn More
Cyphenothrin, Honeywell Fluka™
Brand: Honeywell Fluka R1737-100MG
Additional Details : Weight : 0.10000kg
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Cyphenothrin | |
≤ 100% | |
375.46 g/mol | |
100 mg |
39515-40-7 | |
(RS)-a-cyano-3-phenoxybenzyl (1 RS,3 RS;1 RS,3 SR)-2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropanecarboxylate | |
Yellow | |
130°C |