missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ CyQUANT™ Direct Cell Proliferation Assay

CyQUANT™ Direct Cell Proliferation Assay

£373.00 - £2430.00

Specifications

Color Green
Culture Environment Suspension Cell Culture, Adherent Cell Culture
Detection Method Fluorescence
For Use With (Application) Proliferation Assay
For Use With (Equipment) Microplate Reader, Spectrophotometer, Microscope, HTS Reader
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10422185
View Documents Promotion Details
Molecular Probes™
C35011
1 Kit(s)
£373.00
1 set
Please to purchase this item. Need a web account? Register with us today!
10585675
View Documents Promotion Details
Molecular Probes™
C35012
1 Kit(s)
£2430.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Experience a more convenient workflow for your high-throughput cytotoxicity and proliferation screening projects with the CyQUANT™ Direct Cell Proliferation Assay–no washes, cell lysis, temperature equilibrations, or radioactivity required. DNA-based assays have been shown to be among the most sensitive cell health indicators and data concordance with metabolically based assays is excellent. The CyQUANT™ Direct assay is a highly sensitive and robust method to assess cell growth, viability, or compound toxicity in a range of applications, from high throughput screening to bioproduction.

Features of the CyQUANT™ Direct Cell Proliferation Assay include:

  • More convenient workflow for high-throughput screening–no washes, cell lysis, temperature equilibrations, or radioactivity required
  • Rapid protocol, high sensitivity and dynamic range
  • Measures independent of metabolic state of cells on standard fluorescent plate readers
  • Multiplex assays using a spectrally distinct fluorescent or a luminescent readout

    Rapid and convenient, with a large dynamic range
    The CyQUANT™ Direct assay is a fluorescence-based proliferation and cytotoxicity assay for microplate readers with a linear detection range from less than 100 to 20,000 cells per well in most cell types. The no-wash, homogenous format and fast add-mix-read protocol makes the CyQUANT™ Direct assay ideal for HTS applications. The assay can be completed in one hour, with no washes, no cell lysis, or temperature equilibrations required. The signal is stable for several hours, affording additional work flow convenience.

    How it works
    The CyQUANT™ Direct assay is based on a cell-permeant DNA-binding dye in combination with a masking dye reagent. Cellular DNA is highly regulated and estimates of cell numbers based on DNA are very accurate. The masking dye blocks staining of dead cells or cells with compromised cell membranes resulting in only healthy cells being stained. The CyQUANT™ Direct assay, therefore, measures proliferation as well as membrane integrity, another measure of cell health. Users of metabolically based cell health assays can readily compare historical data with results from the CyQUANT™ Direct assay, as concordance assays based on metabolism or energy (ATP) has been shown to be excellent. Furthermore, it is possible to multiplex with spectrally distinct fluorescent probes or luminescence assay kits because cells are not lysed. Furthermore, the fluorescent signal allows the user to switch between HTS and HCS plate readers if multiplex assays require different platforms.
  • Specifications

    Specifications

    Green
    Fluorescence
    Microplate Reader, Spectrophotometer, Microscope, HTS Reader
    Other Label(s) or Dye(s)
    96-well plate, 1536-well plate, 384-well plate
    CyQUANT™, Molecular Probes™
    Room Temperature
    Suspension Cell Culture, Adherent Cell Culture
    Proliferation Assay
    Cell Proliferation Assay
    Visible
    60 min
    1 Kit(s)
    Videos
    SDS
    Documents

    Documents

    Product Certifications

    For Research Use Only. Not for use in diagnostic procedures.