missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin B Mouse anti-Human, Clone: 2G9B8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66812-1-IG-150UL
This item is not returnable.
View return policy
Description
Cystatin A (also designated STF1, STFA, stefin A or cystatin AS) and cystatin B (also designated PME, CST6, STFB, CPI-B, stefin B and liver thiol proteinase inhibitor) are thiol protease inhibitors that form complexes with Papain and the cathepsins B, H and L. Cystatin A, a cytoplasmic protein, is one of the precursor proteins of the cornified cell envelope in keratinocytes and plays a role in epidermal development and maintece. Cystatin B protects against intracellular proteases leaking out of lysosomes and is primarily expressed in heart, liver and kidney.Specifications
Cystatin B | |
Monoclonal | |
1.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P04080 | |
CSTB | |
Cystatin B Fusion Protein Ag21291 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
2G9B8 | |
Unconjugated | |
CSTB | |
CPI B, CST6, CSTB, Cystatin B, cystatin B (stefin B), EPM1, PME, Stefin B, STFB | |
Mouse | |
Protein G | |
RUO | |
1476 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction