missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Cystatin SA Antibody Pair [HRP]
Rabbit Monoclonal Antibody Pair
£826.11
Specifications
Assay Range | 4.7 pg/mL |
---|---|
Assay Sensitivity | 4.69-300 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Product Type | Antibody Pairs |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18243526
|
Novus Biologicals™
NBP2-79323-15PLATES |
15 Plates |
£826.11
Pack of 15 |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Cystatin SA.Specifications
4.7 pg/mL | |
Sandwich ELISA | |
Antibody Pairs | |
Human | |
cystatin 2, cystatin S5, cystatin SA, Cystatin-2, Cystatin-S5, cystatin-SA, cysteine-proteinase inhibitor, MGC71924, salivary cysteine (thiol) protease inhibitor | |
CST2 | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 0.3 ng/mL is recommended. |
4.69-300 pg/mL | |
HRP | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Cystatin SA. | |
Sandwich ELISA | |
1470 | |
Rabbit | |
RUO | |
Cystatin SA |