missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Cystatin SA Antibody Pair [HRP]

Rabbit Monoclonal Antibody Pair

Brand:  Novus Biologicals™ NBP2-79323-5Plates

 View more versions of this product

Product Code. 18625321

  • £341.00 / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Cystatin SA.
Specifications

Specifications

4.7 pg/mL
Sandwich ELISA
Antibody Pairs
Human
cystatin 2, cystatin S5, cystatin SA, Cystatin-2, Cystatin-S5, cystatin-SA, cysteine-proteinase inhibitor, MGC71924, salivary cysteine (thiol) protease inhibitor
CST2
  • Detection Antibody: 0.25 mg/mL rabbit anti-Cystatin SA monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % glycerol, pH 7.4).
  • Capture Antibody: 1.0 mg/mL of rabbit anti-Cystatin SA monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 15 ng of recombinant Cystatin SA.
  • RUO
    Cystatin SA
    4.69-300 pg/mL
    HRP
    This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Cystatin SA.
    Sandwich ELISA
    1470
    Rabbit
    5 Plates
    Storage is content dependent.
    1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 ug/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20°C to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 0.3 ng/mL is recommended.
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers