missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytarabine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mg
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
Cytarabine is a nucleoside analog that acts as an antileukemic agent
Cytarabine becomes cytotoxic following conversion to its triphosphorylated form, which can be incorporated into DNA primers by DNA polymerase alpha
Can induce S phase cell cycle arrest and apoptosis in leukemic and neuronal cells
Applications
Has been used to study chemotherapeutic regimens in laboratory studies of leukemia
Useful for laboratory studies of cytarabine resistance mechanisms
At nanomolar concentrations, cytarabine has been seen to induce mitochondrial apoptosis in leukemic cells through upregulation of multiple Bcl-2 family members
It can induce neuronal cell death via inducing γ-H2AX phosphorylation in histones, in the absence of double-stranded DNA breaks
Specifications
Specifications
Melting Point | 220.0°C |
Assay Percent Range | 97.5% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 1 g |
Merck Index | 152781 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in dmso, ethanol |
Formula Weight | 243.22 |
Percent Purity | 98% |
Chemical Name or Material | Cytarabine |
Safety and Handling
Product Identifier
- Cytosine beta-D-arabinofuranoside
- Warning
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Respiratory sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H361d-Suspected of damaging the unborn child.
- H319-Causes serious eye irritation.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'