Learn More
Cytarabine, 98%, Thermo Scientific Chemicals
Cytarabine, CAS # 147-94-4, is an inhibitor of DNA synthesis that is used as an anticancer and antiviral agent.
£25.90 - £410.00
Chemical Identifiers
CAS | 147-94-4 |
---|---|
Molecular Formula | C9H13N3O5 |
Molecular Weight (g/mol) | 243.22 |
InChI Key | UHDGCWIWMRVCDJ-CCXZUQQUSA-N |
Synonym | cytarabine, cytosine arabinoside, ara-c, arabinocytidine, cytosar, arabinosylcytosine, aracytidine, cytarabin, cytarabinoside, tarabine |
PubChem CID | 6253 |
ChEBI | CHEBI:28680 |
IUPAC Name | 4-amino-1-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one |
SMILES | C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15244774
|
Thermo Scientific Acros
449561000 |
100 mg | Glass bottle |
£25.90
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15224774
|
Thermo Scientific Acros
449560010 |
1 g | Glass bottle |
£121.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15234774
|
Thermo Scientific Acros
449560050 |
5 g | Glass bottle |
£410.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
147-94-4 | |
243.22 | |
cytarabine, cytosine arabinoside, ara-c, arabinocytidine, cytosar, arabinosylcytosine, aracytidine, cytarabin, cytarabinoside, tarabine | |
CHEBI:28680 | |
C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)O |
C9H13N3O5 | |
UHDGCWIWMRVCDJ-CCXZUQQUSA-N | |
6253 | |
4-amino-1-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one |
Specifications
147-94-4 | |
97.5% min. (HPLC) | |
152781 | |
Solubility in water: soluble. Other solubilities: soluble in dmso, ethanol | |
C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)O | |
243.22 | |
CHEBI:28680 | |
98% |
220.0°C | |
C9H13N3O5 | |
cytarabine, cytosine arabinoside, ara-c, arabinocytidine, cytosar, arabinosylcytosine, aracytidine, cytarabin, cytarabinoside, tarabine | |
UHDGCWIWMRVCDJ-CCXZUQQUSA-N | |
4-amino-1-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one | |
6253 | |
243.22 | |
Cytarabine |
Safety and Handling
Causes serious eye irritation, Suspected of causing genetic defects, Suspected of damaging fertility or the unborn child, May cause an allergic skin reaction
Serious eye damage/eye irritation (category 2), Germ cell mutagenicity (category 2), Reproductive toxicity (category 2), Skin sensitisation (category 1)
missing translation for 'einecsNumber' : 205-705-9
- Cytosine beta-D-arabinofuranoside
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Respiratory sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H361d-Suspected of damaging the unborn child.
- H319-Causes serious eye irritation.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only