Learn More
Thermo Scientific Chemicals Cytidine-5'-diphosphocholine sodium salt
Description
Cytidine-5'-diphosphocholine sodium salt hydrate acts as a neuroprotective agent in situations of hypoxia and ischemia. It is used as biosynthetic intermediate of membrane phospholipids. It is a cerebral vasodilator and inhibits PLA2 activation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Cytidine-5′-diphosphocholine sodium salt hydrate acts as a neuroprotective agent in situations of hypoxia and ischemia. It is used as biosynthetic intermediate of membrane phospholipids. It is a cerebral vasodilator and inhibits PLA2 activation.
Solubility
Soluble in water (100 mg/ml).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Color | White |
| Quantity | 50 g |
| Beilstein | 4110040 |
| Solubility Information | Soluble in water (100mg/ml). |
| Formula Weight | 510.31 (Anhydrous) |
| Physical Form | Powder |
| Chemical Name or Material | Cytidine-5'-diphosphocholine sodium salt hydrate |
RUO – Research Use Only
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.