Learn More
Invitrogen™ CytKick™ Max Autosampler, refurbished
The CytKick Max Autosampler Refurbished is for use with the Attune NxT Acoustic Focusing Cytometer. It enables rapid processing of multiple samples by automatically analyzing samples from 96- and 384-well plates (standard and deep), as well as some custom plate types and microfuge tube racks.
Brand: Invitrogen™ A42973R
Additional Details : Weight : 15.00000kg
Description
This is a refurbished version of the CytKick Max Autosampler, which is for use with the Attune NxT Acoustic Focusing Cytometer. It enables rapid processing of multiple samples by automatically analyzing samples from 96- and 384-well plates (standard and deep), as well as some custom plate types and microfuge tube racks.Specifications
Flow Cytometry Analyzer | |
CytKick™ |