Learn More
Invitrogen™ CytKick™ Max Extended Cooling Adaptor
The CytKick Max Extended Cooling Adaptor is designed for use with the CytKick Max Autosampler and the Attune Acoustic Focusing Cytometer
Brand: Invitrogen™ 100063482
1187.50 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
- The CytKick Max Extended Cooling Adaptor is designed for use with the CytKick Max Autosampler and the Attune Acoustic Focusing Cytometer.
- This cooling block maintains samples at 2-15°C for 95-100 minutes while they are being analyzed on the instrument.
- The cooling adaptor is brought to temperature by placing it in a refrigerator overnight or in a freezer for a minimum of two hours prior to use.
- If this longer cooling time is not required, please see the CytKick Max Universal Cooling Adaptor, which cools samples for ∼65 minutes.
Specifications
CytKick™ | |
Room Temperature | |
Cooling Adapter |
1 adapter | |
Attune Acoustic Focusing Cytometer |