missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Cyto-Plex™ Carboxylated Particles
Shop All Thermo Scientific Products
Click to view available options
Diameter (Metric):
4 μm
5 μm
Quantity:
1 mL
5 mL
Level:
Level 1
Level 10
Level 11
Level 12
Level 2
Level 3
Level 4
Level 5
Level 6
Level 7
Level 8
Level 9
Unit Size:
1mL
5mL
Descripción
Save time when measuring analytes with Thermo Scientific™ Cyto-Plex™ Carboxylated Particles, open platform particles for suspension array analysis.
Suspension array analysis is a powerful technique enabling the simultaneous detection and quantitation of multiple analytes with a single sample. Using color coded microspheres with the appropriate dyes, these tests can now be carried out using commonly available flow cytometers.
- Cyto-Plex™ microspheres consist of a highly uniform polystyrene particle with fluorescent intensities which are completely separated from each other
- Carboxylated particles provide different sizes and levels of fluorescent intensities for analysis of multiple analytes
- Use of a single diameter particle for all dye levels saves time by only requiring the development and optimization of one bead chemistry; multiple diameters can be combined to increase the number of analytes measured in one test
- High-density binding sites and low non-specific binding enable coupling with a wide variety of antibodies, nucleic acids and other biomolecules
- Microspheres have a maximum emission at 700 nm and can be excited with either 488 nm or 633 nm lasers
- Emission can be collected in either the PE-Cy5 or APC channels
- Since there is little or no emission in the FITC and PE channels, probes utilizing either of these dyes can be effectively used as reporters
Recommended for:
- Open platform microspheres for suspension array analysis in flow cytometry
Especificaciones
Especificaciones
| Additive | Sodium Azide Preservative, 0.05% Tween-20 |
| Content And Storage | Refrigerate when not in use; do not freeze. Store upright and keep bottle tightly sealed. Mix product by gentle inversion by hand or vortex mixer. |
| Contents | Polystyrene microspheres in water |
| Description | Cyto-Plex Carboxylated Particles, 4µm |
| Color | Red |
| Material | Polystyrene with Copolymer Grafted Surface |
| Product Type | Carboxylated Particle |
| Dye Type | Firefli™ Fluorescent Red (635/700nm) |
| Includes | Material Safety Data Sheet (MSDS) and Package Insert Sheet |
| Level | Level 2 |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido