missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cyto-Plex™ Carboxylated Particles

Save time when measuring analytes with Thermo Scientific™ Cyto-Plex™ Carboxylated Particles, open platform particles for suspension array analysis.

Brand:  Thermo Scientific™ FM5CR06B

 View more versions of this product

Product Code. 11838353

Alert:

Contains sodium azide.

This item is not returnable. View return policy

Description

Description

Save time when measuring analytes with Thermo Scientific™ Cyto-Plex™ Carboxylated Particles, open platform particles for suspension array analysis.

Suspension array analysis is a powerful technique enabling the simultaneous detection and quantitation of multiple analytes with a single sample. Using color coded microspheres with the appropriate dyes, these tests can now be carried out using commonly available flow cytometers.
  • Cyto-Plex™ microspheres consist of a highly uniform polystyrene particle with fluorescent intensities which are completely separated from each other
  • Carboxylated particles provide different sizes and levels of fluorescent intensities for analysis of multiple analytes
  • Use of a single diameter particle for all dye levels saves time by only requiring the development and optimization of one bead chemistry; multiple diameters can be combined to increase the number of analytes measured in one test
  • High-density binding sites and low non-specific binding enable coupling with a wide variety of antibodies, nucleic acids and other biomolecules
  • Microspheres have a maximum emission at 700 nm and can be excited with either 488 nm or 633 nm lasers
  • Emission can be collected in either the PE-Cy5 or APC channels
  • Since there is little or no emission in the FITC and PE channels, probes utilizing either of these dyes can be effectively used as reporters

Recommended for:

  • Open platform microspheres for suspension array analysis in flow cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sodium Azide Preservative, 0.05% Tween-20
Polystyrene microspheres in water
Red
Carboxylated Particle
Material Safety Data Sheet (MSDS) and Package Insert Sheet
1.06g/cm³
Product stable for a minimum of 12 months
≤2% CV
5 μm
Refrigerate when not in use; do not freeze. Store upright and keep bottle tightly sealed. Mix product by gentle inversion by hand or vortex mixer.
Cyto-Plex Carboxylated Particles, 5µm
Polystyrene with Copolymer Grafted Surface
Firefli™ Fluorescent Red (635/700nm)
Level 6
5 mL
Carboxylate-Modified
1 or 5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number269856 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberFM5CR06B
Lot Number273919_1125 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberFM5CR06B
Lot Number276276_0126 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberFM5CR06B
Lot Number279806_0526 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberFM5CR06B
Lot Number280536_0526 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberFM5CR06B
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Cyto-Plex™ Carboxylated Particles > 5μm; Intensity: Level 6; 5mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.