Learn More
Cytiva Cytodex 3 Gamma Microcarriers
The microcarrier of choice for cells that are difficult to culture in vitro - particularly for cells with an epithelial-like morphology.
£1137.00 - £37000.00
Specifications
For Use With (Application) | Applications include the production of large quantities of cells, viruses and recombinant cell products (e.g. interferon, enzymes, nucleic acids, hormones), studies on cell adhesion, diff erentiation and cell function, perfusion column culture systems, microscopy studies, harvesting mitotic cells, isolation of cells, membrane studies, storage and transport of cells, assays involving celltransfer and studies on uptage of labelled compounds |
---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15747207
|
Cytiva
17548801 |
30 g |
£1137.00
30g |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15767207
|
Cytiva
17548803 |
3 kg |
£37000.00
3kg |
Estimated Shipment: 22-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15757207
|
Cytiva
17548802 |
300 g |
£5338.00
300g |
Estimated Shipment: 22-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Cytodex 3 Gamma is designed by chemically coupling of a thin layer of denatured porcine collagen type 1 to the cross-linked dextran matrix.
- As the collagen surface layer can be digested by a variety of proteolytic enzymes, harvest of such cells from the microcarriers can be performed with maintained high cell viability and membrane integrity.
- This feature of Cytodex 3 Gamma is beneficial in the development of successful serial subcultivation protocols for scaling up culture volumes.
Specifications
Applications include the production of large quantities of cells, viruses and recombinant cell products (e.g. interferon, enzymes, nucleic acids, hormones), studies on cell adhesion, diff erentiation and cell function, perfusion column culture systems, microscopy studies, harvesting mitotic cells, isolation of cells, membrane studies, storage and transport of cells, assays involving celltransfer and studies on uptage of labelled compounds |