missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 16 Mouse anti-Human, Clone: 2H4D8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66802-1-IG-150UL
This item is not returnable.
View return policy
Description
KRT16 is a member of the keratin gene family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. Most of the type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains and are clustered in a region of chromosome 17q12-q21. This keratin has been coexpressed with keratin 14 in a number of epithelial tissues, including esophagus, tongue, and hair follicles. Mutations in this gene are associated with type 1 pachyonychia congenita, non-epidermolytic palmoplantar keratoderma and unilateral palmoplantar verrucous nevus.Specifications
Cytokeratin 16 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P08779 | |
KRT16 | |
Cytokeratin 16 Fusion Protein Ag11240 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
2H4D8 | |
Unconjugated | |
KRT16 | |
CK 16, CK16, Cytokeratin 16, K16, K1CP, keratin 16, KRT16, KRT16A, NEPPK | |
Mouse | |
Protein G | |
RUO | |
3868 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction