missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 17 Monoclonal Antibody (E3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513539
This item is not returnable.
View return policy
Description
MA5-13539 targets Cytokeratin 17 in IF and IHC (P) applications and shows reactivity with Human and Rat samples. The MA5-13539 immunogen is keratin preparation from rat enterocytes.
Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between approximately40 kDa and approximately68 kDa and isoelectric pH between 4. 9 - 7. 8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.Specifications
Cytokeratin 17 | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with 0.09% sodium azide | |
Q04695, Q6IFU8 | |
KRT17 | |
Keratin preparation from rat enterocytes | |
RUO | |
287702, 3872 | |
4° C | |
Liquid |
Immunohistochemistry (Paraffin) | |
E3 | |
Unconjugated | |
KRT17 | |
39.1; CK17; CK-17; cytokeratin-17; hard keratin; K17; Ka17; keratin 17; keratin 17, type I; keratin complex 1, acidic, gene 17; keratin, type I cytoskeletal 17; Keratin-17; Krt1-17; Krt17; krt17 {ECO:0000250; PC; PC2; PCHC1; Type I keratin Ka17; UniProtKB:Q04695} | |
Mouse | |
500 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG2b κ |