missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 17 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17516-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specifically against KRT17.
This gene encodes the type I intermediate filament chain keratin 17, expressed in nail bed, hair follicle, sebaceous glands, and other epidermal appendages. Mutations in this gene lead to Jackson-Lawler type pachyonychia congenita and steatocystoma multiplex.Specifications
Cytokeratin 17 | |
Polyclonal | |
Unconjugated | |
KRT17 | |
39.1, CK 17, Cytokeratin 17, K17, keratin 17, KRT17, PC, PC2, PCHC1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16667, 3872 | |
-20°C | |
Liquid |
Flow Cytometry, Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q04695, Q9QWL7 | |
Krt17 | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction