Learn More
Cytokeratin 17 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17516-1-AP-150UL
 View more versions of this product
Product Code. 16847424
  • £349.00 / 150µL
Explore more special offers
Description
Description
This antibody is specifically against KRT17.
This gene encodes the type I intermediate filament chain keratin 17, expressed in nail bed, hair follicle, sebaceous glands, and other epidermal appendages. Mutations in this gene lead to Jackson-Lawler type pachyonychia congenita and steatocystoma multiplex.
Specifications
Specifications
Cytokeratin 17
Polyclonal
Unconjugated
KRT17
39.1, CK 17, Cytokeratin 17, K17, keratin 17, KRT17, PC, PC2, PCHC1
Rabbit
Antigen Affinity Chromatography
RUO
16667, 3872
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q04695, Q9QWL7
Krt17
Peptide
150 μL
Primary
Human, Mouse