missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 5 Mouse anti-Human, Clone: 1A1C5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66727-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by the KRT5 gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This type II cytokeratin is specifically expressed in the basal layer of the epidermis with family member KRT14. Mutations in these genes have been associated with a complex of diseases termed epidermolysis bullosa simplex. The type II cytokeratins are clustered in a region of chromosome 12q12-q13.Specifications
Cytokeratin 5 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P13647 | |
KRT5 | |
Cytokeratin 5 Fusion Protein Ag24184 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1A1C5 | |
Unconjugated | |
KRT5 | |
58 kDa cytokeratin, CK 5, CK5, Cytokeratin 5, K5, keratin 5, KRT5, KRT5A, Type II keratin Kb5 | |
Mouse | |
Protein G | |
RUO | |
3852 | |
-20°C | |
Liquid |