missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Cytokeratin 8/18 Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 8/18 Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-743-S0

 View more versions of this product

Product Code. 12662227

  • £114.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Keratin 8 belongs to the type B (basic) subfamily of high molecular weight keratins and exists in combination with cytokeratin 18 (type A; acidic subfamily of low molecular weight keratins).

Host Species: Mouse

Clone: 5D3

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Immunogen: Cytokeratins purified from breast cancer MCF-7 cells

Molecular Weight: 52.5 kDa (Cytokeratin 8) and 45 kDa (Cytokeratin 18)

Positive Control: Skin

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Cytokeratin 8/18 Ab-1
Monoclonal
Unconjugated
Cytokeratins purified from breast cancer MCF-7 cells
IVD
Primary
Concentrated
Immunohistochemistry (Paraffin)
5D3
Mouse
100 μL
Cytoskeletal Protein
Human
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.