missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytomegalovirus gB Late Antigen Polyclonal Antibody, Biotin, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA173025
This item is not returnable.
View return policy
Description
The PA1-73025 antibody reacts with Cytomegalovirus AD 169. PA1-73025 has been successfully used in Immunohistochemical, ELISA, Immunofluorescence and Western blot applications. The PA1-73025 antibody was raised against purified virions of strain AD 169 (gB antigen).
Cytomegalovirus is an enveloped double stranded DNA virus which in humans (HCMV) is commonly known as human herpesvirus 5 (HHV-5). Glycoprotein B (gB) is the main antigenic protein within the envelope of HCMV. It is required for HCMV mediated membrane fusion along with the gH/gL complex. The unprocessed precursor of HCMV gB is 906 amino acids in length, with a MW of approximately 100kDa.Specifications
Cytomegalovirus gB Late Antigen | |
Polyclonal | |
Biotin | |
CMV | |
Purified virions of strain AD 169 (gB antigen). | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
4-5 mg/mL | |
PBS with 0.1% sodium azide, pH 7.2 | |
Goat | |
IgG fraction | |
RUO | |
Virus | |
Antibody | |
IgG |