missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Cytopore 2

Brand:  Cytiva 17-1271-01

Product Code. 11300002

  • £5030.00 / 20g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cytopore 2 Microcarriers
Hydrophilic DEAE exchanger, positive charge
230 μm, Spherical
Principally designed for use in suspension culture systems for growth of adherent diploid cells requiring a higher surface charge, and the production of recombinant proteins
Cross-linked cotton cellulose
20 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva Cytopore 2 > Quantity: 20g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.