missing translation for 'onlineSavingsMsg'
Learn More

CytoScint ES, MP Biomedicals™

Product Code. p-3208358 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 gal.
4 x 1 gal.
Unit Size:
15.14 L
3.79 L
This item is not returnable. View return policy

Product Code. 15702252

Brand: MP Biomedicals 0188245301

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Liquid Scintillation Cocktails; CytoScint ES; MP Biomedical; 4 x 1 gal.; Biodegradable

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Liquid Scintillation Cocktails
Quantity 1 gal.
compliance-icons compliance-icons
Product Identifier
  • cytoscint ES
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : phenylxylylethane, linear alkylbenzene,ethoxylated nonylphenol, 2-methoxyethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.