missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
CAS | 71-30-7 |
---|---|
Molecular Formula | C4H5N3O |
Molecular Weight (g/mol) | 111.10 |
MDL Number | MFCD00006034 |
InChI Key | OPTASPLRGRRNAP-UHFFFAOYSA-N |
Synonym | cytosine, 4-amino-2-hydroxypyrimidine, cytosinimine, 2 1h-pyrimidinone, 4-amino, 4-amino-2 1h-pyrimidinone, 4-aminopyrimidin-2 1h-one, 6-aminopyrimidin-2 1h-one, 2 1h-pyrimidinone, 6-amino, cyt, 4-amino-2-oxo-1,2-dihydropyrimidine |
PubChem CID | 597 |
ChEBI | CHEBI:16040 |
SMILES | NC1=CC=NC(=O)N1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10134080
|
Thermo Scientific Acros
161760050 |
5 g | Glass bottle |
£53.97
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10442845
|
Thermo Scientific Acros
161760250 |
25 g | Glass bottle |
£172.86
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15109213
|
Thermo Scientific Acros
161761000 |
100 g | Glass bottle |
£558.78
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
71-30-7 | |
111.10 | |
OPTASPLRGRRNAP-UHFFFAOYSA-N | |
597 | |
NC1=CC=NC(=O)N1 |
C4H5N3O | |
MFCD00006034 | |
cytosine, 4-amino-2-hydroxypyrimidine, cytosinimine, 2 1h-pyrimidinone, 4-amino, 4-amino-2 1h-pyrimidinone, 4-aminopyrimidin-2 1h-one, 6-aminopyrimidin-2 1h-one, 2 1h-pyrimidinone, 6-amino, cyt, 4-amino-2-oxo-1,2-dihydropyrimidine | |
CHEBI:16040 |
Specifications
71-30-7 | |
White to Yellow | |
Authentic | |
C4H5N3O | |
24,314 | |
cytosine, 4-amino-2-hydroxypyrimidine, cytosinimine, 2 1h-pyrimidinone, 4-amino, 4-amino-2 1h-pyrimidinone, 4-aminopyrimidin-2 1h-one, 6-aminopyrimidin-2 1h-one, 2 1h-pyrimidinone, 6-amino, cyt, 4-amino-2-oxo-1,2-dihydropyrimidine | |
98% min. (lambda max. 275nm,pH 1) molar absorptivity 10400+/-2%,on dry basis | |
NC1=CC=NC(=O)N1 | |
111.10 | |
CHEBI:16040 | |
99+% | |
Cytosine |
>300.0°C | |
0.5% max. (105°C, 5 hrs) | |
99% min. (HPLC) | |
MFCD00006034 | |
15,2792 | |
Solubility in water: soluble. Other solubilities: soluble in alkalines and acids,slightly soluble in ethanol,chloroform,,insoluble in ether and hydrocarbons | |
OPTASPLRGRRNAP-UHFFFAOYSA-N | |
6-amino-1H-pyrimidin-2-one | |
597 | |
111.1 | |
Crystalline Powder |
Safety and Handling
missing translation for 'einecsNumber' : 200-749-5
Product Identifier
- Cytosine
RUO – Research Use Only