Learn More
Epredia™ Cytoslide™ Microscope Slides
Improve processing and visualization of small-volume specimens with these cytoslides, for use with Epredia™ Shandon™ EZ Single Cytofunnels™.
Brand:  Epredia™ 5991056
98.75 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.
 View more versions of this product
Product Code. 12026689
  • £159.00 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Uncoated slides available with circle on front or back
  • Available in boxes of 100 slides
Specifications
Specifications
Glass
Single Cytoslide
Plain
Coated, circle on back