Learn More
Epredia™ Cytoslide™ Microscope Slides
Improve processing and visualization of small-volume specimens with these cytoslides, for use with Epredia™ Shandon™ EZ Single Cytofunnels™.
Brand: Epredia™ 5991051
65.55 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
- Uncoated slides available with circle on front or back
- Available in cases of 100 slides
Specifications
Single Cytoslide | |
Plain | |
Shandon™ Single Cytoslides™; Noncoated, circle on back |