missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytosolic Sulfotransferase 2B1/SULT2B1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-16997-100UL
This item is not returnable.
View return policy
Description
Cytosolic Sulfotransferase 2B1/SULT2B1 Polyclonal antibody specifically detects Cytosolic Sulfotransferase 2B1/SULT2B1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Cytosolic Sulfotransferase 2B1/SULT2B1 | |
Polyclonal | |
Western Blot 0.04 to 0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Alcohol sulfotransferase, EC 2.8.2, EC 2.8.2.2, HSST2sulfotransferase family cytosolic 2B member 1, Hydroxysteroid sulfotransferase 2, ST2B1, Sulfotransferase 2B1, sulfotransferase family, cytosolic, 2B, member 1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: KAKVIYMGRNPRDVVVSLYHYSKIAGQLKDPGTPDQFLRDFLKGEVQFGSWFDH | |
100 μg | |
metabolism | |
6820 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction