missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(-)-2-Phenylglycine ethyl ester hydrochloride, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
D-(-)-2-Phenylglycine ethyl ester hydrochloride is used in pharmaceutical intermediates, in the combination of dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-(-)-2-Phenylglycine ethyl ester hydrochloride is used in pharmaceutical intermediates, in the combination of dyestuff.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
D-(-)-2-Phenylglycine ethyl ester hydrochloride is used in pharmaceutical intermediates, in the combination of dyestuff.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
Melting Point | ∽190°C (decomposition) |
Beilstein | 5775455 |
Solubility Information | Soluble in water. |
Quantity | 50 g |
Formula Weight | 215.68 |
Optical Rotation | −94° (c=1 in Water) |
Percent Purity | ≥98% |
Chemical Name or Material | D-(-)-2-Phenylglycine ethyl ester hydrochloride |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction