Learn More
Thermo Scientific™ D.A.R.T.'S™ (Disposable Automation Research Tips), with Extended Length
Achieve ultimate performance in automated pipetting with Thermo Scientific™ D.A.R.T.'S™ Extended Length tips — superior quality with the flexibility to choose tips for any application.
£143.00 - £2395.00
Specifications
Certifications/Compliance | ISO 9001 and ISO 13485 |
---|---|
Clean Claims | Free of RNase, DNase, DNA, ATP, and endotoxin |
Tip Style | Automation |
Disposable | Single-use |
Product Code | Brand | Volume (Metric) | Filtered | Sterility | Packaging | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Filtered | Sterility | Packaging | Price | Quantity & Availability | ||
10029552
|
Thermo Scientific™
5418 |
30 μL | Filtered | Non-sterile | 20 x 384 tips |
£2395.00
Pack of 7680 |
Estimated Shipment: 24-12-2024
Log in to see stock. |
||
15313032
|
Thermo Scientific™
5502-11 |
25 μL | Filtered | Sterile |
£143.00
Pack of 10 |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||
10621354
|
Thermo Scientific™
5562-11 |
220 μL | Filtered | Sterile |
£227.00
Pack of 1920 |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||
10243673
|
Thermo Scientific™
5508 |
30 μL | Filtered | Sterile | 20 x 96 tips |
£483.00
Pack of 1920 |
Estimated Shipment: 27-11-2024
Log in to see stock. |
||
11597459
|
Thermo Scientific™
5538 |
300 μL | Filtered | Sterile | 20 x 96 tips |
£580.00
Pack of 1920 |
Estimated Shipment: 24-12-2024
Log in to see stock. |
||
10304883
|
Thermo Scientific™
5548 |
300 μL | Filtered | Sterile | 20 x 96 tips |
£510.00
Pack of 1920 |
Estimated Shipment: 24-12-2024
Log in to see stock. |
||
10314883
|
Thermo Scientific™
5561 |
220 μL | Non-Filtered | Non-sterile | 20 x 96 tips |
£205.00
Pack of 1920 |
Estimated Shipment: 17-12-2024
Log in to see stock. |
||
10325213
|
Thermo Scientific™
5416 |
30 μL | Non-Filtered | Non-sterile | 20 x 384 tips |
£1665.00
Pack of 7680 |
Estimated Shipment: 17-12-2024
Log in to see stock. |
||
10168981
|
Thermo Scientific™
5536 |
300 μL | Non-Filtered | Non-sterile | 20 x 96 tips |
£327.00
Pack of 1920 |
Estimated Shipment: 15-11-2024
Log in to see stock. |
||
10405963
|
Thermo Scientific™
5546 |
300 μL | Non-Filtered | Non-sterile | 20 x 96 tips |
£333.00
Pack of 1920 |
Estimated Shipment: 17-12-2024
Log in to see stock. |
||
10690024
|
Thermo Scientific™
5417 |
30 μL | Non-Filtered | Sterile | 20 x 384 tips |
£1690.00
Pack of 7680 |
Estimated Shipment: 24-12-2024
Log in to see stock. |
||
10283623
|
Thermo Scientific™
5507 |
30 μL | Non-Filtered | Sterile | 20 x 96 tips |
£303.00
Pack of 1920 |
Estimated Shipment: 06-12-2024
Log in to see stock. |
||
11868423
|
Thermo Scientific™
5537 |
300 μL | Non-Filtered | Sterile | 20 x 96 tips |
£363.00
Pack of 1920 |
In Stock
Log in to see stock. |
||
10394873
|
Thermo Scientific™
5547 |
300 μL | Non-Filtered | Sterile | 20 x 96 tips |
£347.00
Pack of 1920 |
In Stock
Log in to see stock. |
||
Description
Perform pipetting more effectively with Thermo Scientific™ Matrix™ SerialMate™ Extended Length Pipette Tips, for use with the Matrix SerialMate™ Pipetter. Unlike conventional automation tips, D.A.R.T.'s™ are packaged in disposable rigid magazines and seal onto the pipetting heads by being pressed against a silicone pad. This definitive seal minimizes volume displacement, ensures tip straightness and enables consistent dispensing for improved well access. A small internal orifice provides small droplet formation resulting in optimal accuracy and precision.
Features:- Stepped-body design minimizes volume displacement and ensures tip straightness for better well access.
- Face seal design ensures consistent tip-to-tip height for precise low volume dispensing
- Highly polished molds and cores ensure low liquid retention.
- Small internal tip orifice provides better small volume droplet formation, resulting in optimal accuracy and precision.
- Sterile, sterile filtered and wide-bore tip options are available
- Tips are thoroughly inspected for proper formation and straightness
- Unique face seal design ensures consistent tip-to-tip height for precise low volume dispensing.
- Entire assembly can be quickly removed and replaced eliminating extra handling of the tips, which can cause damage or contamination.
Compatible with: Thermo Scientific™ Matrix™ Hydra™ DT, Matrix™ PlateMate™ 2x2, Matrix™ PlateMate™ 2x3, PlateMate™ Plus Systems and Versette.
Specifications
ISO 9001 and ISO 13485 | |
Free of RNase, DNase, DNA, ATP, and endotoxin | |
Automation | |
Single-use |