missing translation for 'onlineSavingsMsg'
Learn More

D-Alanine ethyl ester hydrochloride, 95%, Thermo Scientific Chemicals

£37.80 - £141.00

Chemical Identifiers

CAS 6331-09-5
MDL Number MFCD00190725
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15429268
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H62791.06
5 g
£37.80
5g
Please to purchase this item. Need a web account? Register with us today!
15439268
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H62791.14
25 g
£141.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

D-Alanine ethyl ester hydrochloride is used as organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Alanine ethyl ester hydrochloride is used as organic intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

6331-09-5
MFCD00190725

Specifications

6331-09-5
MFCD00190725
CYHRNDXVTAWKMO-UHFFFAOYNA-N
ethyl (2R)-2-aminopropanoate;hydrochloride
10236019
95%
C5H11ClNO2
d-alanine ethyl ester hydrochloride, r-ethyl 2-aminopropanoate hydrochloride, h-d-ala-oet hcl, r-ethyl 2-amino-propionate hcl, ethyl 2r-2-aminopropanoate hydrochloride, h-ala-oblz. tos, h-d-ala-oet. hcl, 3-alanine ethyl ester hydrochloride, r-ethyl 2-amino-propionate hydrochloride
CCOC(=O)C(C)N.Cl
152.60
153.61
D-Alanine ethyl ester hydrochloride
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only