missing translation for 'onlineSavingsMsg'
Learn More

D-Alanine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 15438988
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

D-Alanine methyl ester hydrochloride is used as building block for the preparation of peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Alanine methyl ester hydrochloride is used as building block for the preparation of peptides.

Solubility
Soluble in Water 100 mg/ml.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14316-06-4
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.58
MDL Number MFCD00066141
InChI Key IYUKFAFDFHZKPI-UHFFFAOYNA-N
Synonym d-alanine methyl ester hydrochloride, methyl d-alaninate hydrochloride, h-d-ala-ome.hcl, methyl d-alaninate hcl, methyl 2r-2-aminopropanoate hydrochloride, h-d-ala-ome hcl, r-methyl 2-aminopropanoate hydrochloride, d-alanine methylester hydrochloride, d-alanine, methyl ester, hydrochloride, pubchem10885
PubChem CID 11182647
IUPAC Name methyl (2R)-2-aminopropanoate;hydrochloride
SMILES [H+].[Cl-].COC(=O)C(C)N

Specifications

Melting Point 108°C to 110°C
Color White
Beilstein 4235975
Solubility Information Soluble in Water 100mg/ml.
Quantity 100 g
Formula Weight 139.58
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material D-Alanine methyl ester hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.