missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-allo-Threonine, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
They are found as constituents of an increasing group of biologically active peptides. Stereocontrolled syntheses of chiral and racemic key intermediates to thienamycin from d-allo-threonine and trans-crotonic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
They are found as constituents of an increasing group of biologically active peptides. Stereocontrolled syntheses of chiral and racemic key intermediates to thienamycin from d-allo-threonine and trans-crotonic acid.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
They are found as constituents of an increasing group of biologically active peptides. Stereocontrolled syntheses of chiral and racemic key intermediates to thienamycin from d-allo-threonine and trans-crotonic acid.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 268°C (decomposition) |
Beilstein | 1721644 |
Solubility Information | Soluble in water. |
Formula Weight | 119.12 |
Percent Purity | 99% |
Quantity | 250 mg |
Optical Rotation | −8.7° (c=2 in Water) |
Chemical Name or Material | D-allo-Threonine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'