missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ D-amino acid oxidase (Porcine)
Used for Enzyme
Brand: Abnova™ P5306.5mg
Additional Details : Weight : 0.00010kg
Description
#REF!Specifications
P00371 | |
397134 | |
D-amino acid oxidase (Porcine) | |
5 mg | |
RUO | |
DAO1 | |
D-amino acid oxidase Assay; Keto acid preparation; Oxidation reduction studies; Separation of L-amino acids from racemic mixtures; FAD determination; D-alanine determination. | |
Pig | |
Native protein chromatographically purified from Porcine Kidney |
Enzyme Activity | |
∽39kDa | |
Native protein chromatographically purified from Porcine Kidney | |
Store at 4°C. | |
DAAO/DAMOX/DAO | |
DAO1 | |
≥2 units per mg dry weight. One Unit oxidizes 1 umole of D-alanine per minute at 37°C, pH 8.3. | |
None | |
Lyophilized |