missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Asparagine monohydrate
Click to view available options
Quantity:
1 g
5 g
25 g
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifica
Melting Point | 280.0°C |
Molecular Formula | C4H8N2O3·H2O |
Linear Formula | H2NCOCH2CH(NH2)CO2H·H2O |
Beilstein | 04, 471 |
Merck Index | 14, 837 |
Solubility Information | Solubility aq. soln.: 35 g/L (20°C). Other solubilities: soluble in acids and alkalies, practically insoluble in methanol, ethanol, ether, and benzene |
Formula Weight | 150.13 |
Quantity | 25 g |
Chemical Name or Material | D-Asparagine monohydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'