missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Biotin, ≥97.5%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Color | White |
Assay Percent Range | ≥97.5% |
Quantity | 5 g |
Formula Weight | 244.3 |
Physical Form | Crystalline Powder |
Chemical Name or Material | D-Biotin |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
D-Biotin, ≥97.5%, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction