Learn More
D-(+)-Biotin, 98+%, Thermo Scientific Chemicals
Description
Biotin is a form of vitamin B that helps the body to break down fats and carbohydrates. It is used as an alternative medicine to treat or prevent biotin deficiency, which is caused by malnutrition, rapid weight loss, long-term tube feeding and other medical conditions. It is an essential vitamin which is important for amino acid and energy metabolism.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in hot water, dimethyl sulfoxide, alcohol and benzene.
Notes
Store in cool place. Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 224°C to 226°C |
Assay Percent Range | ≥98% |
Quantity | 1 g |
Beilstein | 86838 |
Sensitivity | Light sensitive |
Merck Index | 14,1231 |
Solubility Information | Soluble in hot water,dimethyl sulfoxide,alcohol and benzene. |
Formula Weight | 244.32 |
Percent Purity | ≥98% |
Chemical Name or Material | D-(+)-Biotin |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N24L039 | Certificate of Analysis | 26/02/2025 | A14207.06,A14207.14,A14207.03,A14207.09 |
10253363 | Certificate of Analysis | 11/12/2024 | A14207.06,A14207.03,A14207.14,A14207.09 |
10252481 | Certificate of Analysis | 18/09/2024 | A14207.06,A14207.03,A14207.14,A14207.09 |
10251062 | Certificate of Analysis | 30/08/2024 | A14207.06,A14207.03,A14207.14,A14207.09 |
10252240 | Certificate of Analysis | 29/08/2024 | A14207.06,A14207.03,A14207.14,A14207.09 |
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Days
0Hours
8Minutes
53Seconds
38
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.