missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Calcium pantothenate, 98%
15.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 190°C |
Color | White |
Assay Percent Range | 8.2 to 8.6% (Ca) On dried substance (Complexometry) |
Infrared Spectrum | Authentic |
Linear Formula | [HOCH2C(CH3)2CHOHCONHCH2CH2COO]2Ca |
Loss on Drying | 5.0% max. (105°C, 3 hrs) |
Merck Index | 15, 7118 |
Packaging | Glass Bottle |
Solubility Information | Solubility in water: 350g/L (20°C). Other solubilities: soluble in glycerine,very slightly soluble in ethanol,insoluble in ether,chloroform |
Specific Rotation | + 25 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'