missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Calcium pantothenate, 98%

Product Code. 10712964
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-08-6
Molecular Formula C18H32CaN2O10
Molecular Weight (g/mol) 476.54
MDL Number MFCD00002766
InChI Key FAPWYRCQGJNNSJ-DXHDTSSINA-L
Synonym calcium pantothenate, d-pantothenic acid hemicalcium salt, d-pantothenic acid, calcium salt, pantothenic acid, calcium salt, d, r-n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-beta-alanine calcium salt, beta-alanine, n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-, calcium salt, r
PubChem CID 131847364
IUPAC Name calcium;3-[[(2R)-2,4-dihydroxy-3,3-dimethylbutanoyl]amino]propanoic acid
SMILES [Ca++].CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O.CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O

Specifications

Melting Point 190°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 8.2 to 8.6% (Ca) On dried substance (Complexometry)
Loss on Drying 5.0% max. (105°C, 3 hrs)
Packaging Plastic Bottle
Linear Formula [HOCH2C(CH3)2CHOHCONHCH2CH2COO]2Ca
Specific Rotation + 25
Merck Index 15, 7118
Solubility Information Solubility in water: 350g/L (20°C). Other solubilities: soluble in glycerine,very slightly soluble in ethanol,insoluble in ether,chloroform
Specific Rotation Condition +25.00 (20.00°C c=5,H2O)
Quantity 500 g
Formula Weight 476.52
Percent Purity 98%
Physical Form Powder
Chemical Name or Material D-Calcium pantothenate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.