missing translation for 'onlineSavingsMsg'
Learn More

D-CPP-ene, Tocris Bioscience™

Product Code. 18065832
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent competitive NMDA antagonist

Potent and competitive NMDA antagonist (Ki = 40 nM). Centrally active following systemic administration.

Specifications

CAS 117414-74-1
Molecular Formula C8H15N2O5P
Quantity 10 mg
Synonym midafotel, d-cpp-ene, unii-7lyu6zf84g, sdz-eaa 494, 7lyu6zf84g, sdz-eaa-494, --r-4-e-3-phosphonoallyl-2-piperazinecarboxylic acid, midafotel inn, d-cppene, d-4-2e-3-phosphono-2-propenyl-2-piperazinecarboxylic acid
InChI Key VZXMZMJSGLFKQI-ABVWVHJUSA-N
SMILES C1CN(CC(N1)C(=O)O)CC=CP(=O)(O)O
IUPAC Name (2R)-4-[(E)-3-phosphonoprop-2-enyl]piperazine-2-carboxylic acid
Molecular Weight (g/mol) 250.191
PubChem CID 6435801
Formula Weight 250.19
Percent Purity >98%
Chemical Name or Material D-CPP-ene
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'