missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Cysteine, 99%
Click to view available options
Quantity:
1 g
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 921-01-7 |
Color | White |
Assay Percent Range | 99% |
Molecular Formula | C3H7NO2S |
Infrared Spectrum | Authentic |
Linear Formula | HSCH2CH(NH2)CO2H |
MDL Number | MFCD00066461 |
Packaging | Glass Bottle |
Synonym | d-cysteine, d-cystein, 2s-2-amino-3-sulfanylpropanoic acid, d-zystein, cysteine, d, s-2-amino-3-mercaptopropanoic acid, d-amino-3-mercaptopropionic acid, d-cys, h-d-cys-oh, cysteine |
InChI Key | XUJNEKJLAYXESH-RUOZJJLTNA-N |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'