missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals D-erythro-Sphingosine 1-phosphate, 96% Product Code.: 10019580

Thermo Scientific Chemicals D-erythro-Sphingosine 1-phosphate, 96%

Product Code. 10019580
1 mg
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy

Product Code. 10019580

Brand: Thermo Scientific Chemicals 328520010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26993-30-6 | C18H38NO5P | 379.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Especificaciones

CAS 26993-30-6
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 95% min
Molecular Formula C18H38NO5P
MDL Number MFCD00274457
Quantity 1 mg
Synonym sphingosine 1-phosphate, sphingosine-1-phosphate, c18-sphingosine 1-phosphate, d-erythro-sphingosine-1-phosphate, sphing-4-enine 1-phosphate, d-erythro-sphingosine 1-phosphate, sphingosine 1-phosphic acid, s1p, sphing-4-enine-1-phosphate, 2s,3r,4e-2-amino-3-hydroxyoctadec-4-en-1-yl dihydrogen phosphate
InChI Key DUYSYHSSBDVJSM-KRWOKUGFSA-N
SMILES CCCCCCCCCCCCCC=CC(C(COP(=O)(O)O)N)O
IUPAC Name [(E,2S,3R)-2-amino-3-hydroxyoctadec-4-enyl] dihydrogen phosphate
Molecular Weight (g/mol) 379.5
PubChem CID 5283560
ChEBI CHEBI:37550
Formula Weight 379.5
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material D-erythro-Sphingosine 1-phosphate
Mostrar más Mostrar menos

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467510 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number328520010
Lot NumberA0452371 Certificate TypeCertificate of Analysis Date03/05/2023 Catalog Number328520010
Lot NumberA0440828 Certificate TypeCertificate of Analysis Date05/04/2022 Catalog Number328520010
Lot NumberA0413902 Certificate TypeCertificate of Analysis Date13/05/2021 Catalog Number328520010
compliance-icons
Nombre del producto
  • D-erythro-Sphingosine 1-phosphate
Palabra de advertencia
  • Warning
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Consejos de prudencia
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Thermo Scientific Chemicals D-erythro-Sphingosine 1-phosphate, 96% >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado