missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-erythro-Sphingosine, 99+%, synthetical

Product Code. 10594141
Click to view available options
Quantity:
50 mg
250 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-78-4
Molecular Formula C18H37NO2
Molecular Weight (g/mol) 299.5
MDL Number MFCD00036751
InChI Key WWUZIQQURGPMPG-KRWOKUGFSA-N
Synonym sphingosine, d-erythro-sphingosine, 4-sphingenine, d-sphingosine, sphing-4-enine, cerebroside, sphingenine, 4-trans-sphingenine, trans-d-erythro-2-amino-4-octadecene-1,3-diol, 2s,3r,4e-2-amino-4-octadecene-1,3-diol
PubChem CID 5280335
ChEBI CHEBI:16393
IUPAC Name (E,2S,3R)-2-aminooctadec-4-ene-1,3-diol
SMILES CCCCCCCCCCCCCC=CC(C(CO)N)O

Specifications

Melting Point 81.0°C to 82.0°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 250 mg
Merck Index 15, 8874
Solubility Information Solubility in water: insoluble in water. Other solubilities: insoluble in acetone, soluble in chloroform and dichloromethane
Formula Weight 299.5
Percent Purity 99+%
Physical Form Powder or Waxy Solid
Chemical Name or Material D-erythro-Sphingosine, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'