missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-erythro-Sphingosine, 99+%, synthetical
£149.00 - £564.00
Chemical Identifiers
CAS | 123-78-4 |
---|---|
Molecular Formula | C18H37NO2 |
Molecular Weight (g/mol) | 299.5 |
MDL Number | MFCD00036751 |
InChI Key | WWUZIQQURGPMPG-KRWOKUGFSA-N |
Synonym | sphingosine, d-erythro-sphingosine, 4-sphingenine, d-sphingosine, sphing-4-enine, cerebroside, sphingenine, 4-trans-sphingenine, trans-d-erythro-2-amino-4-octadecene-1,3-diol, 2s,3r,4e-2-amino-4-octadecene-1,3-diol |
PubChem CID | 5280335 |
ChEBI | CHEBI:16393 |
IUPAC Name | (E,2S,3R)-2-aminooctadec-4-ene-1,3-diol |
SMILES | CCCCCCCCCCCCCC=CC(C(CO)N)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10524871
|
Thermo Scientific Chemicals
331530500 |
50 mg | Glass bottle |
£149.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10594141
|
Thermo Scientific Chemicals
331532500 |
250 mg | Glass bottle |
£564.00
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
123-78-4 | |
299.5 | |
WWUZIQQURGPMPG-KRWOKUGFSA-N | |
5280335 | |
(E,2S,3R)-2-aminooctadec-4-ene-1,3-diol |
C18H37NO2 | |
MFCD00036751 | |
sphingosine, d-erythro-sphingosine, 4-sphingenine, d-sphingosine, sphing-4-enine, cerebroside, sphingenine, 4-trans-sphingenine, trans-d-erythro-2-amino-4-octadecene-1,3-diol, 2s,3r,4e-2-amino-4-octadecene-1,3-diol | |
CHEBI:16393 | |
CCCCCCCCCCCCCC=CC(C(CO)N)O |
Specifications
123-78-4 | |
White to Cream | |
99+% | |
MFCD00036751 | |
sphingosine, d-erythro-sphingosine, 4-sphingenine, d-sphingosine, sphing-4-enine, cerebroside, sphingenine, 4-trans-sphingenine, trans-d-erythro-2-amino-4-octadecene-1,3-diol, 2s,3r,4e-2-amino-4-octadecene-1,3-diol | |
WWUZIQQURGPMPG-KRWOKUGFSA-N | |
(E,2S,3R)-2-aminooctadec-4-ene-1,3-diol | |
5280335 | |
299.5 | |
Powder or Waxy Solid |
81.0°C to 82.0°C | |
Authentic | |
C18H37NO2 | |
15, 8874 | |
Solubility in water: insoluble in water. Other solubilities: insoluble in acetone, soluble in chloroform and dichloromethane | |
CCCCCCCCCCCCCC=CC(C(CO)N)O | |
299.5 | |
CHEBI:16393 | |
99+% | |
D-erythro-Sphingosine, 99+% |
Safety and Handling
missing translation for 'einecsNumber' : 204-651-3
RUO – Research Use Only