Learn More
Thermo Scientific Chemicals D-Fructose-6-phosphate disodium salt, 95%
Description
D-Fructose-6-phosphate disodium salt is a sugar intermediate of the glycolytic pathway that may be used to help identify, differentiate and characterize enzymes such as phosphofructokinase(s), pyrophosphate-dependent fructose-6-phosphate 1-phosphotransferase(s), D-fructose-6-phosphate aldolase(s), glutamine:fructose-6-phosphate amidotransferase(s) and glucosamine-6P synthase(s).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
D-Fructose-6-phosphate disodium salt is a sugar intermediate of the glycolytic pathway that may be used to help identify, differentiate and characterize enzymes such as phosphofructokinase(s), pyrophosphate-dependent fructose-6-phosphate 1-phosphotransferase(s), D-fructose-6-phosphate aldolase(s), glutamine:fructose-6-phosphate amidotransferase(s) and glucosamine-6P synthase(s).
Solubility
Soluble in water (100 mg/ml).
Notes
Store at -20°C. Incompatible with heat and oxidizing agents.
Specifications
Specifications
Color | White |
Quantity | 1 g |
Solubility Information | Soluble in water (100mg/ml). |
Formula Weight | 304.1 |
Percent Purity | 95% |
Physical Form | Powder |
Chemical Name or Material | D-Fructose-6-phosphate disodium salt |
Safety and Handling
- D-Fructose-6-phosphate disodium salt
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : D-Fructose-6-phosphate disodium salt
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.